Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032826.4 | NP_116215.1 | 2 | 471 | ||
XM_011516645.2 | XP_011514947.1 | 2 | 239 | |||
GenBank mRNA | AB052892.1 | - | 2 | 447 | ||
AJ315497.1 | - | 2 | 303 | |||
AJ315498.1 | - | 2 | 293 | |||
AJ971941.1 | - | 2 | 147 | |||
AK027603.1 | - | 2 | 311 | |||
AK027726.1 | - | 1 | 29 | |||
AK075368.1 | - | 2 | 276 | |||
AK222874.1 | - | 2 | 320 | |||
AK307621.1 | - | 2 | 299 | |||
BC008413.1 | - | 2 | 292 | 4333393 | ||
DA208815.1 | - | 2 | 471 |