Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032827.6 | NP_116216.2 | 3 | 2093 | ||
XM_011533139.1 | XP_011531441.1 | 4 | 2151 | |||
GenBank mRNA | AK027614.1 | - | 3 | 1886 | ||
AK074681.1 | - | 3 | 1985 | |||
AK096049.1 | - | 3 | 1440 | |||
BC021207.2 | - | 3 | 1984 | 4109799 | ||
BC094832.1 | - | 3 | 1763 | 6292270 |