Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032839.2 | NP_116228.1 | 3 | 622 | ||
GenBank mRNA | AK027690.1 | - | 3 | 616 | ||
AK075158.1 | - | 3 | 539 | |||
AK291176.1 | - | 3 | 606 | |||
BC022204.1 | - | 3 | 880 | |||
BC033738.1 | - | 2 | 498 | |||
BC039821.1 | - | 3 | 593 | 6062886 |