Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032854.3 | NP_116243.2 | 10 | 1744 | ||
XM_005258047.3 | XP_005258104.1 | 11 | 2026 | |||
XM_005258048.4 | XP_005258105.2 | 11 | 1963 | |||
XM_005258056.4 | XP_005258113.1 | 6 | 1840 | |||
XM_011525385.2 | XP_011523687.1 | 11 | 1960 | |||
XM_011525386.2 | XP_011523688.1 | 10 | 1858 | |||
XM_011525387.2 | XP_011523689.1 | 10 | 1840 | |||
XM_011525390.2 | XP_011523692.1 | 7 | 1706 | |||
XM_011525391.2 | XP_011523693.1 | 7 | 1684 | |||
XM_017025235.1 | XP_016880724.1 | 10 | 1907 | |||
GenBank mRNA | AF193039.1 | - | 5 | 942 | ||
AF447884.1 | - | 7 | 1725 | |||
AF447885.1 | - | 5 | 1408 | |||
AK027777.1 | - | 5 | 1413 | |||
AK092430.1 | - | 8 | 1656 | |||
AK094683.1 | - | 6 | 2566 | |||
AK096431.1 | - | 10 | 1649 | |||
AL833952.1 | - | 8 | 3136 | |||
BC048209.1 | - | 7 | 1155 | |||
BC064514.1 | - | 7 | 1155 | 3678185 |