Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286378.1 | NP_001273307.1 | 3 | 198 | ||
NM_032858.2 | NP_116247.1 | 2 | 425 | |||
XM_006711583.1 | XP_006711646.1 | 3 | 229 | |||
XM_011510067.1 | XP_011508369.1 | 2 | 400 | |||
XM_011510068.1 | XP_011508370.1 | 3 | 191 | |||
XM_017002602.1 | XP_016858091.1 | 4 | 566 | |||
XM_017002603.1 | XP_016858092.1 | 3 | 228 | |||
GenBank mRNA | AK027810.1 | - | 2 | 232 | ||
AK131066.1 | - | 2 | 235 | |||
BC028595.2 | - | 2 | 205 | 5270252 | ||
DQ076156.2 | - | 2 | 234 | |||
GU383114.1 | - | 3 | 198 |