Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PNISR SIRNA Assays ›
Gene Symbol:
PNISR
Gene Name:
PNN interacting serine and arginine rich protein
Gene Aliases:
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2
Chromosome Location:
Chr.6: 99398051 - 99425345 on Build GRCh38
UniGene ID:
Hs.520287
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
34947
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PNISR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322405.1
NP_001309334.1
4
421
NM_001322406.1
NP_001309335.1
5
537
NM_001322408.1
NP_001309337.1
3
341
NM_001322410.1
NP_001309339.1
4
425
NM_001322412.1
NP_001309341.1
4
421
NM_001322413.1
NP_001309342.1
3
345
NM_001322414.1
NP_001309343.1
4
425
NM_001322415.1
NP_001309344.1
3
345
NM_001322416.1
NP_001309345.1
4
425
NM_001322417.1
NP_001309346.1
4
425
NM_001322418.1
NP_001309347.1
3
341
NM_001322419.1
NP_001309348.1
4
425
NM_015491.2
NP_056306.1
3
345
NM_032870.3
NP_116259.2
4
425
NR_136326.1
3
345
XM_005266912.3
XP_005266969.2
3
304
XM_017010710.1
XP_016866199.1
4
422
XM_017010711.1
XP_016866200.1
3
308
XM_017010712.1
XP_016866201.1
4
422
XM_017010713.1
XP_016866202.1
4
418
GenBank mRNA
AF314184.1
-
2
215
AF314186.1
-
4
363
AJ420570.1
-
3
2006
AK027658.1
-
4
388
AK027759.1
-
5
364
AK027898.1
-
4
381
AK074628.1
-
3
308
AK291155.1
-
2
194
BC007791.2
-
4
358
BC012780.2
-
4
367
BC019074.2
-
4
334
BC042850.1
-
5
534
BC052638.1
-
4
363
BC064413.1
-
4
359
BC064640.1
-
4
338
BC107700.1
-
4
376
BX648520.1
-
3
272
DA208838.1
-
4
411
DA326969.1
-
4
425
DB186090.1
-
5
477
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001025274.
This siRNA also targets the following transcripts in Mouse: NM_025669, AK083021, BC141031, AK160288, AK077040, and others.
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-phbv4:80/100.66.79.66:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline