Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286739.1 | NP_001273668.1 | 5, 6 | 587 | ||
NM_001286740.1 | NP_001273669.1 | 4, 5 | 452 | |||
NM_001286741.1 | NP_001273670.1 | 4, 5 | 942 | |||
NM_001286742.1 | NP_001273671.1 | 5, 6 | 684 | |||
NM_032907.4 | NP_116296.1 | 4, 5 | 573 | |||
NM_201265.1 | NP_957717.1 | 4, 5 | 502 | |||
XM_011522124.2 | XP_011520426.1 | 5, 6 | 789 | |||
GenBank mRNA | AY037166.1 | - | 4, 5 | 476 | ||
BC007913.2 | - | 4, 5 | 856 | 4302762 | ||
BC030055.1 | - | 5, 6 | 550 | 4873512 | ||
BC033919.1 | - | 5, 6 | 591 | 4904699 | ||
BG704085.1 | - | 4, 5 | 499 | |||
BI551787.1 | - | 4, 5 | 559 | |||
BI552240.1 | - | 4, 5 | 508 |