Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032839.2 | NP_116228.1 | 9 | 2053 | ||
GenBank mRNA | AK027690.1 | - | 9 | 2047 | ||
AK075158.1 | - | 10 | 2065 | |||
BC022204.1 | - | 9 | 2311 | |||
BC039821.1 | - | 9 | 2024 | 6062886 | ||
BP377383.1 | - | 1 | 208 |