Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128211.1 | NP_001121683.1 | 6 | 919 | ||
NM_032869.3 | NP_116258.2 | 6 | 1019 | |||
XM_017013910.1 | XP_016869399.1 | 7 | 1195 | |||
XM_017013911.1 | XP_016869400.1 | 7 | 965 | |||
GenBank mRNA | AF283301.1 | - | 6 | 1140 | ||
AF283302.1 | - | 6 | 877 | |||
AF521133.1 | - | 6 | 919 | |||
AK027897.1 | - | 6 | 971 | |||
AK301278.1 | - | 7 | 1035 | |||
AK307663.1 | - | 6 | 920 | |||
AK311583.1 | - | 7 | 971 | |||
AK315006.1 | - | 6 | 988 | |||
AL832317.1 | - | 3 | 327 | |||
BC000967.2 | - | 6 | 964 | 3449287 | ||
BC031258.1 | - | 7 | 1109 | 5272186 | ||
BC043406.1 | - | 6 | 984 | 6059975 |