Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032871.3 | NP_116260.2 | 11 | 3263 | ||
NM_152222.1 | NP_689408.1 | 11 | 3201 | |||
XM_005274356.2 | XP_005274413.1 | 6 | 3364 | |||
XM_006718714.2 | XP_006718777.1 | 9 | 2828 | |||
XM_011545305.1 | XP_011543607.1 | 10 | 3086 |