Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033031.2 | NP_149020.2 | 3 | 263 | ||
NM_033670.2 | NP_391990.1 | 3 | 263 | |||
XM_017029913.1 | XP_016885402.1 | 4 | 447 | |||
XM_017029914.1 | XP_016885403.1 | 4 | 377 | |||
XM_017029915.1 | XP_016885404.1 | 5 | 407 | |||
XM_017029916.1 | XP_016885405.1 | 4 | 447 | |||
XM_017029917.1 | XP_016885406.1 | 4 | 447 | |||
XM_017029918.1 | XP_016885407.1 | 4 | 447 | |||
GenBank mRNA | AJ314764.1 | - | 3 | 267 | ||
AJ314765.1 | - | 3 | 267 | |||
AJ314766.1 | - | 3 | 267 | |||
AJ416458.1 | - | 3 | 279 | |||
AK292454.1 | - | 4 | 436 |