Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033083.6 | NP_149074.3 | 2 | 373 | ||
XM_005265518.1 | XP_005265575.1 | 2 | 373 | |||
XM_011534165.1 | XP_011532467.1 | 2 | 522 | |||
XM_011534166.1 | XP_011532468.1 | 2 | 517 | |||
GenBank mRNA | AF272973.1 | - | 2 | 304 | ||
AK304697.1 | - | 2 | 332 | |||
AL698790.1 | - | 2 | 373 | |||
AW292089.1 | - | 2 | 78 | |||
BC041329.2 | - | 2 | 315 | 5284115 | ||
BI459315.1 | - | 2 | 582 |