Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 3, 4 | 389 | ||
GenBank mRNA | AF268032.1 | - | 3, 4 | 350 | ||
AF423421.1 | - | 3, 4 | 323 | |||
AJ347750.1 | - | 3, 4 | 323 | |||
AK098246.1 | - | 3, 4 | 359 | |||
AK126506.1 | - | 1, 2 | 782 | |||
AK315105.1 | - | 3, 4 | 359 | |||
AL831950.1 | - | 2, 3 | 237 | |||
BC036447.2 | - | 3, 4 | 359 | 4830913 | ||
DC348657.1 | - | 3, 4 | 390 |