Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033105.4 | NP_149096.2 | 3 | 383 | ||
XM_011517618.2 | XP_011515920.1 | 3 | 315 | |||
XM_011517620.2 | XP_011515922.1 | 3 | 376 | |||
GenBank mRNA | AF368276.1 | - | 3 | 361 | ||
AL133660.1 | - | 3 | 325 | |||
BC016742.2 | - | 3 | 380 | 4830917 | ||
BG701336.1 | - | 3 | 399 |