Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033131.3 | NP_149122.1 | 2 | 285 | ||
XM_011544319.2 | XP_011542621.1 | 2 | 256 | |||
GenBank mRNA | AB060284.1 | - | 2 | 183 | ||
AK056278.1 | - | 2 | 256 | |||
BC103921.1 | - | 2 | 225 | 40007188 | ||
BC103922.2 | - | 2 | 225 | 40007189 | ||
BC103923.1 | - | 2 | 225 | 40007191 | ||
DA742143.1 | - | 2 | 285 |