Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033133.4 | NP_149124.3 | 2 | 638 | ||
XM_006721701.3 | XP_006721764.1 | 2 | 662 | |||
XM_011524340.2 | XP_011522642.1 | 2 | 623 | |||
GenBank mRNA | AK026876.1 | - | 2 | 595 | ||
AK097833.1 | - | 392 | ||||
AK098384.1 | - | 2 | 584 | |||
BC001362.2 | - | 2 | 575 | 3051112 | ||
BC006392.2 | - | 2 | 597 | 4126696 | ||
BC011046.1 | - | 2 | 559 | 4179083 | ||
BC028040.1 | - | 2 | 599 | 5248370 | ||
M19650.1 | - | 1 | 447 |