Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012101.3 | NP_036233.2 | 1 | 882 | ||
XM_011542729.1 | XP_011541031.1 | 1 | 906 | |||
XM_011542732.1 | XP_011541034.1 | 1 | 906 | |||
GenBank mRNA | AF230388.1 | - | 1 | 882 | ||
AF230389.1 | - | 1 | 882 | |||
AK122720.1 | - | 2 | 698 | |||
AK223090.1 | - | 1 | 840 | |||
AK299472.1 | - | 1 | 239 | |||
BC010644.1 | - | 1 | 828 | 3855016 | ||
BC017352.2 | - | 1 | 821 | 5013158 | ||
L24203.1 | - | 1 | 882 |