Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135733.1 | NP_001129205.1 | 3 | 545 | ||
NM_033285.3 | NP_150601.1 | 3 | 545 | |||
XM_011517386.2 | XP_011515688.1 | 3 | 397 | |||
GenBank mRNA | AB017926.1 | - | 2 | 200 | ||
AB017927.1 | - | 2 | 200 | |||
AF409114.1 | - | 3 | 517 | |||
AF409115.1 | - | 3 | 517 | |||
AK125880.1 | - | 3 | 524 | |||
AK315071.1 | - | 3 | 487 | |||
AL833652.1 | - | 2 | 125 | |||
BC074813.2 | - | 2 | 193 | 30915317 | ||
BC074868.2 | - | 2 | 193 | 30915467 |