Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB362928.1 | - | 2 | 470 | ||
AF220025.1 | - | 2 | 532 | |||
AF220026.1 | - | 532 | ||||
AF220027.1 | - | 2 | 532 | |||
AF220028.1 | - | 2 | 532 | |||
AF220029.1 | - | 2 | 532 | |||
AK027593.1 | - | 3 | 727 | |||
AY625000.1 | - | 2 | 470 | |||
BC020770.1 | - | 4 | 896 | |||
BC021258.2 | - | 3 | 706 | 4122548 | ||
BX647152.1 | - | 3 | 1253 | |||
DQ288685.1 | - | 2 | 470 | |||
JF928461.1 | - | 2 | 484 |