Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171971.2 | NP_001165442.1 | 5 | 535 | ||
NM_033100.3 | NP_149091.1 | 5 | 535 | |||
XM_011540337.1 | XP_011538639.1 | 4 | 1026 | |||
XM_011540338.1 | XP_011538640.1 | 4 | 1026 | |||
XM_011540339.1 | XP_011538641.1 | 5 | 409 | |||
XM_011540340.2 | XP_011538642.1 | 4 | 1026 | |||
GenBank mRNA | AB053448.1 | - | 5 | 408 | ||
AB593154.1 | - | 5 | 535 | |||
AB593155.1 | - | 5 | 521 | |||
BC038799.1 | - | 5 | 525 | 5492127 |