Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ458465.1 | - | 4 | 386 | ||
AY027528.1 | - | 4 | 382 | |||
BC053660.1 | - | 4 | 369 | 6062060 |