Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033132.3 | NP_149123.2 | 2 | 1846 | ||
GenBank mRNA | AF378304.1 | - | 2 | 1831 |