Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033281.5 | NP_150597.1 | 3 | 222 | ||
GenBank mRNA | AF086034.1 | - | 2 | 53 | ||
AF271777.1 | - | 326 | ||||
BC015966.1 | - | 3 | 212 | 4068931 | ||
BC017900.1 | - | 3 | 152 | 4251462 | ||
BX951861.1 | - | 3 | 228 |