Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033328.2 | NP_201585.1 | 1 | 610 | ||
GenBank mRNA | AB053259.1 | - | 1 | 585 | ||
AK058174.1 | - | 1 | 572 | |||
BC016745.2 | - | 1 | 614 | 4839348 | ||
BG772483.1 | - | 1 | 619 | |||
GQ472212.1 | - | 1 | 614 |