Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284358.1 | NP_001271287.1 | 7 | 867 | ||
NM_001284361.1 | NP_001271290.1 | 10 | 1116 | |||
NM_001284362.1 | NP_001271291.1 | 9 | 1050 | |||
NM_033030.5 | NP_149019.1 | 9 | 963 | |||
NM_197970.2 | NP_932074.1 | 9 | 900 | |||
XM_006712715.3 | XP_006712778.1 | 9 | 1260 | |||
XM_011511692.2 | XP_011509994.1 | 10 | 1326 | |||
XM_011511693.2 | XP_011509995.1 | 10 | 1326 | |||
XM_011511694.2 | XP_011509996.1 | 10 | 933 | |||
GenBank mRNA | AF272858.1 | - | 9 | 908 | ||
AK058099.1 | - | 9 | 963 | |||
AK093453.1 | - | 10 | 1116 | |||
AK097795.1 | - | 9 | 1259 | |||
AK098824.1 | - | 9 | 932 | |||
AK302817.1 | - | 7 | 867 | |||
AM295005.1 | - | 9 | 1050 | |||
BC033674.1 | - | 9 | 915 | 5170732 |