Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033034.2 | NP_149023.2 | 6 | 1158 | ||
NM_033092.2 | NP_149083.2 | 6 | 1158 | |||
NM_033093.2 | NP_149084.2 | 6 | 1010 | |||
XM_005253183.2 | XP_005253240.1 | 6 | 1010 | |||
XM_005253184.2 | XP_005253241.1 | 6 | 1144 | |||
XM_011520427.1 | XP_011518729.1 | 5 | 1121 | |||
XM_017018462.1 | XP_016873951.1 | 6 | 1144 | |||
GenBank mRNA | AB362928.1 | - | 5 | 855 | ||
AF220025.1 | - | 5 | 917 | |||
AF220026.1 | - | 917 | ||||
AF220027.1 | - | 5 | 917 | |||
AF220028.1 | - | 5 | 917 | |||
AF220029.1 | - | 4 | 894 | |||
AK027593.1 | - | 6 | 1112 | |||
AY625000.1 | - | 5 | 855 | |||
BC021258.2 | - | 6 | 1091 | 4122548 | ||
DQ288685.1 | - | 5 | 855 | |||
JF928462.1 | - | 4 | 846 |