Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033083.6 | NP_149074.3 | 3, 4 | 552 | ||
GenBank mRNA | AF272973.1 | - | 3, 4 | 483 | ||
AK312330.1 | - | 3, 4 | 524 | |||
AW292089.1 | - | 3, 4 | 257 | |||
BC041329.2 | - | 3, 4 | 494 | 5284115 |