Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 14 | 1737 | ||
GenBank mRNA | AF423421.1 | - | 14 | 1671 | ||
AJ347750.1 | - | 14 | 1671 | |||
AK095001.1 | - | 5 | 507 | |||
AK098246.1 | - | 14 | 1707 | |||
AK126506.1 | - | 11 | 2005 | |||
AK300775.1 | - | 13 | 1631 | |||
AK315105.1 | - | 14 | 1707 | |||
AL831950.1 | - | 13 | 1585 | |||
BC036447.2 | - | 14 | 1707 | 4830913 |