Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033123.3 | NP_149114.2 | 14 | 1952 | ||
NR_073075.1 | 14 | 1955 | ||||
XM_005253519.2 | XP_005253576.1 | 13 | 2068 | |||
XM_011520883.1 | XP_011519185.1 | 11 | 1358 | |||
XM_017020176.1 | XP_016875665.1 | 13 | 2068 | |||
XM_017020177.1 | XP_016875666.1 | 14 | 1945 | |||
XM_017020178.1 | XP_016875667.1 | 12 | 1918 | |||
XM_017020179.1 | XP_016875668.1 | 13 | 2063 | |||
XM_017020180.1 | XP_016875669.1 | 13 | 1957 | |||
XM_017020181.1 | XP_016875670.1 | 13 | 2022 | |||
XM_017020182.1 | XP_016875671.1 | 9 | 1721 | |||
XM_017020183.1 | XP_016875672.1 | 10 | 1485 | |||
XM_017020184.1 | XP_016875673.1 | 10 | 1413 | |||
XM_017020185.1 | XP_016875674.1 | 9 | 1298 | |||
XM_017020186.1 | XP_016875675.1 | 11 | 1320 | |||
GenBank mRNA | AF532185.1 | - | 14 | 1877 | ||
AK097555.1 | - | 11 | 1469 | |||
AK097725.1 | - | 10 | 1418 | |||
AK292279.1 | - | 14 | 1950 | |||
AK314516.1 | - | 14 | 1909 | |||
AY035866.1 | - | 14 | 1914 | |||
BC125067.1 | - | 10 | 1205 | 40117397 |