Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033125.3 | NP_149116.2 | 2 | 255 | ||
XM_011536204.2 | XP_011534506.1 | 2 | 269 | |||
XM_011536205.2 | XP_011534507.1 | 2 | 227 | |||
XM_011536210.1 | XP_011534512.1 | 2 | 227 | |||
GenBank mRNA | AF268892.1 | - | 2 | 188 | ||
AK304546.1 | - | 2 | 227 | |||
AY145502.1 | - | 2 | 188 | |||
BC012014.1 | - | 2 | 186 | |||
BC037247.1 | - | 2 | 203 | |||
BC047565.1 | - | 2 | 255 | 4838011 | ||
GQ472228.1 | - | 2 | 255 |