Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171157.1 | NP_001164628.1 | 3 | 795 | ||
NM_001322105.1 | NP_001309034.1 | 3 | 795 | |||
NM_033130.4 | NP_149121.2 | 3 | 795 | |||
XM_005259366.4 | XP_005259423.1 | 3 | 623 | |||
GenBank mRNA | AF301007.1 | - | 3 | 690 | ||
AF310233.1 | - | 3 | 610 | |||
AF311905.1 | - | 3 | 795 | |||
AK289900.1 | - | 3 | 581 | |||
AK292025.1 | - | 3 | 698 | |||
AK303717.1 | - | 3 | 634 | |||
AK308982.1 | - | 2 | 319 | |||
BC101725.1 | - | 3 | 660 | 8069231 | ||
BC143497.1 | - | 3 | 660 | 9052005 |