Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135733.1 | NP_001129205.1 | 5 | 2757 | ||
NM_033285.3 | NP_150601.1 | 4 | 2719 | |||
XM_011517386.2 | XP_011515688.1 | 4 | 2571 | |||
GenBank mRNA | AB017926.1 | - | 3 | 2373 | ||
AB017927.1 | - | 4 | 2411 | |||
AF409114.1 | - | 5 | 2729 | |||
AF409115.1 | - | 4 | 2691 | |||
AK123608.1 | - | 1 | 911 | |||
AK125880.1 | - | 5 | 2736 | |||
AL833652.1 | - | 4 | 2336 |