Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMKN SIRNA Assays ›
Gene Symbol:
DMKN
Gene Name:
dermokine
Gene Aliases:
UNQ729, ZD52F10
Chromosome Location:
Chr.19: 35497217 - 35513678 on Build GRCh38
UniGene ID:
Hs.417795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
35341
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMKN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035516.3
NP_001030593.1
6
549
NM_001126056.2
NP_001119528.2
18
1849
NM_001126059.2
NP_001119531.1
12
900
NM_001190347.1
NP_001177276.1
17
1801
NM_001308380.1
NP_001295309.1
13
963
NM_001308383.1
NP_001295312.1
11
870
NM_033317.4
NP_201574.3
16
1882
NR_033746.1
14
990
XM_006723477.1
XP_006723540.1
16
1079
XM_006723484.1
XP_006723547.1
14
989
XM_006723489.1
XP_006723552.1
16
1065
XM_006723493.2
XP_006723556.1
14
970
XM_006723494.2
XP_006723557.1
14
952
XM_006723503.2
XP_006723566.1
16
998
XM_011527494.2
XP_011525796.1
22
2088
XM_011527495.2
XP_011525797.1
23
2123
XM_011527496.2
XP_011525798.1
21
2052
XM_011527497.2
XP_011525799.1
21
2046
XM_011527498.2
XP_011525800.1
22
2081
XM_011527499.2
XP_011525801.1
21
2043
XM_011527500.2
XP_011525802.1
21
2034
XM_011527501.2
XP_011525803.1
21
2028
XM_011527502.2
XP_011525804.1
20
1998
XM_011527503.2
XP_011525805.1
20
1992
XM_011527504.2
XP_011525806.1
20
1974
XM_011527505.2
XP_011525807.1
19
1947
XM_011527506.2
XP_011525808.1
19
1938
XM_011527507.2
XP_011525809.1
18
1902
XM_011527508.2
XP_011525810.1
18
1887
XM_011527509.2
XP_011525811.1
17
1845
XM_011527510.2
XP_011525812.1
17
1842
XM_011527513.1
XP_011525815.1
13
938
XM_011527514.1
XP_011525816.1
12
878
XM_017027475.1
XP_016882964.1
14
983
XM_017027476.1
XP_016882965.1
12
906
XM_017027477.1
XP_016882966.1
12
790
GenBank mRNA
AF086315.1
-
6
540
AK096215.1
-
12
1529
AL832080.1
-
6
550
BC004493.2
-
14
872
BC011886.2
-
14
919
BC035311.1
-
16
1876
4752921
EF158323.1
-
12
881
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cornified envelope assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-wqf8l:80/100.66.76.116:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline