Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033345.3 | NP_203131.1 | 2, 3 | 181 | ||
XM_011510089.2 | XP_011508391.1 | 3, 4 | 1221 | |||
XM_011510091.2 | XP_011508393.1 | 2, 3 | 198 | |||
XM_017002631.1 | XP_016858120.1 | 6, 7 | 671 | |||
XM_017002632.1 | XP_016858121.1 | 4, 5 | 502 | |||
XM_017002633.1 | XP_016858122.1 | 3, 4 | 700 | |||
XM_017002634.1 | XP_016858123.1 | 3, 4 | 379 | |||
GenBank mRNA | BC069718.1 | - | 2, 3 | 131 | 7262591 | |
BX281622.1 | - | 2, 3 | 192 | |||
BX954018.1 | - | 2, 3 | 85 |