Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282902.2 | NP_001269831.1 | 2 | 320 | ||
NM_001282903.2 | NP_001269832.1 | 3 | 809 | |||
NM_014112.4 | NP_054831.2 | 3 | 823 | |||
XM_005251049.2 | XP_005251106.1 | 2 | 638 | |||
XM_006716625.1 | XP_006716688.1 | 3 | 760 | |||
XM_011517264.1 | XP_011515566.1 | 4 | 893 | |||
XM_011517266.2 | XP_011515568.1 | 3 | 628 | |||
XM_011517267.1 | XP_011515569.1 | 3 | 746 | |||
XM_011517268.1 | XP_011515570.1 | 3 | 735 | |||
XM_017013801.1 | XP_016869290.1 | 2 | 4422 | |||
GenBank mRNA | AF183810.1 | - | 3 | 817 | ||
AF264784.1 | - | 2 | 364 | |||
AK304046.1 | - | 2 | 315 | |||
AK309357.1 | - | 2 | 534 | |||
BC125020.1 | - | 3 | 653 | 40073132 | ||
DC339831.1 | - | 2 | 320 |