Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320304.1 | NP_001307233.1 | 4 | 953 | ||
NM_001320306.1 | NP_001307235.1 | 4 | 972 | |||
NM_001320307.1 | NP_001307236.1 | 4 | 999 | |||
NM_001320309.1 | NP_001307238.1 | 3 | 1109 | |||
NM_001320310.1 | NP_001307239.1 | 4 | 986 | |||
NM_001320311.1 | NP_001307240.1 | 4 | 986 | |||
NM_033416.2 | NP_219484.1 | 4 | 959 | |||
GenBank mRNA | AF054996.1 | - | 4 | 774 | ||
AK289878.1 | - | 4 | 287 | |||
AK292345.1 | - | 4 | 287 | |||
AK312808.1 | - | 4 | 287 | |||
AY364245.1 | - | 3 | 251 | |||
BC010042.1 | - | 4 | 268 | 3629513 | ||
DA154765.1 | - | 3 | 441 | |||
DA352603.1 | - | 4 | 297 | |||
DA598011.1 | - | 4 | 316 | |||
DA949366.1 | - | 4 | 301 | |||
DB200539.1 | - | 4 | 293 | |||
DB211328.1 | - | 4 | 281 |