Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033026.1 | NP_001028198.1 | 1, 2 | 379 | ||
NM_033420.3 | NP_219488.1 | 1, 2 | 379 | |||
XM_005259675.2 | XP_005259732.1 | 1, 2 | 406 | |||
XM_005259676.2 | XP_005259733.1 | 1, 2 | 406 | |||
XM_005259677.2 | XP_005259734.1 | 1, 2 | 406 | |||
XM_005259678.2 | XP_005259735.1 | 1, 2 | 406 | |||
XM_017027457.1 | XP_016882946.1 | 1, 2 | 406 | |||
GenBank mRNA | AK095756.1 | - | 1, 2 | 379 | ||
BC008957.2 | - | 1, 2 | 352 | 2964663 | ||
DQ005958.1 | - | 1, 2 | 361 |