Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 3 | 445 | ||
NM_001042633.2 | NP_001036098.1 | 3 | 445 | |||
NM_033421.3 | NP_219489.1 | 3 | 445 | |||
NM_152897.2 | NP_690857.1 | 3 | 445 | |||
XM_011529096.2 | XP_011527398.1 | 3 | 858 | |||
XM_011529098.2 | XP_011527400.1 | 3 | 858 | |||
XM_011529099.2 | XP_011527401.1 | 3 | 858 | |||
XM_017028120.1 | XP_016883609.1 | 3 | 858 | |||
GenBank mRNA | AF395845.1 | - | 3 | 356 | ||
AF523834.1 | - | 3 | 424 | |||
AK091359.1 | - | 1 | 449 | |||
AK095851.1 | - | 2 | 326 | |||
AK293443.1 | - | 3 | 446 | |||
AK297622.1 | - | 2 | 368 | |||
AL591562.1 | - | 3 | 424 | |||
BC012904.1 | - | 3 | 445 | |||
BC019823.1 | - | 3 | 418 | 4453502 | ||
BM925584.1 | - | 3 | 431 | |||
DA053179.1 | - | 3 | 446 | |||
KU178820.1 | - | 3 | 356 | |||
KU178821.1 | - | 3 | 356 |