Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033540.2 | NP_284941.2 | 2 | 228 | ||
XM_005247596.3 | XP_005247653.2 | 2 | 186 | |||
GenBank mRNA | AB043588.1 | - | 1 | 193 | ||
AB043589.1 | - | 2 | 193 | |||
AF329637.1 | - | 1 | 102 | |||
AK000700.1 | - | 2 | 185 | |||
AK314306.1 | - | 2 | 185 | |||
BC020661.1 | - | 2 | 194 | |||
BC040557.1 | - | 2 | 228 | 5270347 |