Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079691.1 | NP_001073159.1 | 5 | 690 | ||
NM_001286459.1 | NP_001273388.1 | 5 | 698 | |||
NM_052818.2 | NP_438169.2 | 5 | 698 | |||
XM_006719890.3 | XP_006719953.1 | 6 | 1060 | |||
XM_011535301.2 | XP_011533603.1 | 6 | 851 | |||
XM_011535302.2 | XP_011533604.1 | 8 | 1132 | |||
XM_011535303.2 | XP_011533605.1 | 5 | 1261 | |||
XM_011535304.2 | XP_011533606.1 | 8 | 1109 | |||
XM_017020836.1 | XP_016876325.1 | 6 | 2251 | |||
XM_017020837.1 | XP_016876326.1 | 4 | 861 | |||
XM_017020838.1 | XP_016876327.1 | 8 | 1192 | |||
XM_017020840.1 | XP_016876329.1 | 8 | 1107 | |||
XM_017020841.1 | XP_016876330.1 | 8 | 1200 | |||
XM_017020842.1 | XP_016876331.1 | 9 | 1238 | |||
GenBank mRNA | AL049785.1 | - | 5 | 654 | ||
AL049786.1 | - | 5 | 654 | |||
AL049787.1 | - | 5 | 646 | |||
AL049788.1 | - | 7 | 712 | |||
BC022188.2 | - | 5 | 636 | |||
BC125050.1 | - | 4 | 367 | |||
BX648509.1 | - | 4 | 3627 | |||
U50527.1 | - | 5 | 641 |