Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052855.3 | NP_443087.1 | 2 | 462 | ||
XM_017025296.1 | XP_016880785.1 | 2 | 209 | |||
GenBank mRNA | AK314010.1 | - | 2 | 209 | ||
BC012978.2 | - | 2 | 454 | 3834331 | ||
CR627413.1 | - | 2 | 234 | |||
EU164539.1 | - | 2 | 454 |