Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 2 | 185 | ||
XM_005265537.4 | XP_005265594.1 | 2 | 198 | |||
XM_006713384.3 | XP_006713447.1 | 2 | 196 | |||
XM_011534214.2 | XP_011532516.1 | 2 | 200 | |||
XM_011534215.2 | XP_011532517.1 | 2 | 200 | |||
XM_017007460.1 | XP_016862949.1 | 2 | 197 | |||
XM_017007461.1 | XP_016862950.1 | 2 | 186 | |||
GenBank mRNA | AJ318099.1 | - | 2 | 169 | ||
BC043595.1 | - | 2 | 178 | 5769824 |