Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172831.1 | NP_001166302.1 | 6 | 694 | ||
NM_001172832.1 | NP_001166303.1 | 4 | 597 | |||
NM_052860.2 | NP_443092.1 | 5 | 639 | |||
XM_011537702.2 | XP_011536004.1 | 6 | 794 | |||
GenBank mRNA | AF395541.1 | - | 5 | 485 | ||
AK095642.1 | - | 2 | 298 | |||
AK096465.1 | - | 5 | 636 | |||
AK300528.1 | - | 6 | 615 | |||
BC117248.1 | - | 5 | 388 | 40125799 | ||
DQ908918.1 | - | 6 | 500 |