Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135911.1 | NP_001129383.1 | 4 | 681 | ||
NM_052880.4 | NP_443112.2 | 4 | 681 | |||
GenBank mRNA | AF528079.1 | - | 4 | 626 | ||
AF528080.1 | - | 4 | 626 | |||
AK093768.1 | - | 1 | 1241 | |||
AK226089.1 | - | 4 | 561 | |||
AK299397.1 | - | 4 | 635 | |||
AK300990.1 | - | 5 | 539 | |||
BC011049.2 | - | 4 | 626 | 4180134 | ||
BC041903.1 | - | 2 | 978 | |||
BE299406.1 | - | 2 | 309 | |||
BQ718653.1 | - | 4 | 659 | |||
CR456340.1 | - | 4 | 521 | |||
GU135609.1 | - | 2 | 305 |