Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052891.2 | NP_443123.1 | 3 | 388 | ||
XM_011509118.1 | XP_011507420.1 | 5 | 817 | |||
XM_011509119.1 | XP_011507421.1 | 4 | 709 | |||
XM_011509120.2 | XP_011507422.1 | 5 | 595 | |||
GenBank mRNA | AY035376.1 | - | 3 | 388 | ||
BC069741.1 | - | 3 | 374 | 7262478 | ||
BC069798.1 | - | 3 | 375 | 7262490 | ||
BC069801.1 | - | 3 | 375 | 7262502 | ||
BC128114.1 | - | 3 | 361 | 40112227 | ||
BC128115.1 | - | 3 | 361 | 40112231 |