Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK11IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052902.3 | NP_443134.3 | 4 | 405 | ||
XM_011510526.1 | XP_011508828.1 | 4 | 385 | |||
XM_011510527.2 | XP_011508829.1 | 4 | 385 | |||
XM_011510529.1 | XP_011508831.1 | 4 | 385 | |||
GenBank mRNA | AB067485.1 | - | 2 | 83 | ||
AF450267.1 | - | 4 | 359 | |||
AK091963.1 | - | 3 | 287 | |||
AK295609.1 | - | 4 | 388 | |||
AK296632.1 | - | 4 | 385 | |||
AK299894.1 | - | 3 | 585 | |||
AK300611.1 | - | 4 | 388 | |||
BC034051.1 | - | 4 | 382 | 4819148 | ||
HY001118.1 | - | 4 | 408 |