Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286250.1 | NP_001273179.1 | 3 | 627 | ||
NM_001286251.1 | NP_001273180.1 | 3 | 627 | |||
NM_001286252.1 | NP_001273181.1 | 3 | 627 | |||
NM_001323252.1 | NP_001310181.1 | 4 | 674 | |||
NM_001323253.1 | NP_001310182.1 | 3 | 627 | |||
NM_001323254.1 | NP_001310183.1 | 3 | 627 | |||
NM_001323255.1 | NP_001310184.1 | 3 | 627 | |||
NM_001323256.1 | NP_001310185.1 | 3 | 627 | |||
NM_001323257.1 | NP_001310186.1 | 3 | 627 | |||
NM_001323258.1 | NP_001310187.1 | 4 | 2371 | |||
NM_001323259.1 | NP_001310188.1 | 3 | 627 | |||
NM_001323260.1 | NP_001310189.1 | 3 | 627 | |||
NM_052904.3 | NP_443136.2 | 3 | 627 | |||
NR_104421.1 | 3 | 627 | ||||
NR_136580.1 | 3 | 627 | ||||
GenBank mRNA | AB067487.1 | - | 1 | 41 | ||
AF351610.1 | - | 3 | 455 | |||
AK055292.1 | - | 3 | 513 | |||
AK295479.1 | - | 3 | 452 | |||
AK297240.1 | - | 3 | 371 | |||
BC031830.1 | - | 2 | 208 | |||
BC071627.1 | - | 3 | 502 | 5245540 | ||
DA296671.1 | - | 3 | 519 | |||
DA332224.1 | - | 4 | 399 |