Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199196.1 | NP_001186125.1 | 5 | 1263 | ||
NM_033415.3 | NP_219483.1 | 4 | 1155 | |||
XM_005260157.3 | XP_005260214.1 | 4 | 1206 | |||
XM_017027484.1 | XP_016882973.1 | 4 | 1130 | |||
XM_017027485.1 | XP_016882974.1 | 6 | 1387 | |||
GenBank mRNA | AK295488.1 | - | 4 | 929 | ||
AK303914.1 | - | 2 | 843 | |||
BC056161.1 | - | 6 | 885 | |||
BC067125.1 | - | 5 | 1032 | 6161112 | ||
BX648486.1 | - | 4 | 1050 | |||
CR749678.1 | - | 1 | 496 |