Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320304.1 | NP_001307233.1 | 4 | 962 | ||
NM_001320306.1 | NP_001307235.1 | 4 | 981 | |||
NM_001320307.1 | NP_001307236.1 | 4 | 1008 | |||
NM_001320309.1 | NP_001307238.1 | 3 | 1118 | |||
NM_001320310.1 | NP_001307239.1 | 4 | 995 | |||
NM_001320311.1 | NP_001307240.1 | 4 | 995 | |||
NM_033416.2 | NP_219484.1 | 4 | 968 | |||
GenBank mRNA | AF054996.1 | - | 4 | 783 | ||
AK289878.1 | - | 4 | 296 | |||
AK292345.1 | - | 4 | 296 | |||
AK312808.1 | - | 4 | 296 | |||
AY364245.1 | - | 3 | 260 | |||
BC010042.1 | - | 4 | 277 | 3629513 | ||
DA154765.1 | - | 3 | 450 | |||
DA352603.1 | - | 4 | 306 | |||
DA598011.1 | - | 4 | 325 | |||
DA949366.1 | - | 4 | 310 | |||
DB200539.1 | - | 4 | 302 | |||
DB211328.1 | - | 4 | 290 |