Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163499.1 | NP_001156971.1 | 4 | 370 | ||
NM_012172.4 | NP_036304.2 | 4 | 520 | |||
NM_033506.2 | NP_277041.1 | 4 | 631 | |||
XM_005250259.3 | XP_005250316.1 | 4 | 805 | |||
XM_011516022.1 | XP_011514324.1 | 4 | 363 | |||
XM_011516023.2 | XP_011514325.1 | 3 | 622 | |||
XM_017011961.1 | XP_016867450.1 | 5 | 2106 | |||
GenBank mRNA | AF174604.1 | - | 4 | 296 | ||
AK292240.1 | - | 4 | 638 | |||
AK301864.1 | - | 4 | 464 | |||
AK302252.1 | - | 4 | 370 | |||
AK310487.1 | - | 4 | 576 | |||
AL136811.1 | - | 4 | 576 | |||
BC067293.1 | - | 4 | 571 | 30383439 | ||
DC397982.1 | - | 4 | 520 |